siRNA Details
| Gene Symbol: Inpp5k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013859.1 | NP_001013881.1 | 12 | 1410 | ||
| GenBank mRNA | BC083572.1 | - | 12 | 1410 | 7112698 | |
| FQ228522.1 | - | 10 | 2319 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Inpp5k| Gene Symbol: Inpp5k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013859.1 | NP_001013881.1 | 12 | 1410 | ||
| GenBank mRNA | BC083572.1 | - | 12 | 1410 | 7112698 | |
| FQ228522.1 | - | 10 | 2319 | |||