siRNA Details
| Gene Symbol: Ift20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105815.1 | NP_001099285.1 | 5 | 650 | ||
| XM_006246935.2 | XP_006246997.1 | 4 | 464 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ift20| Gene Symbol: Ift20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105815.1 | NP_001099285.1 | 5 | 650 | ||
| XM_006246935.2 | XP_006246997.1 | 4 | 464 | |||