siRNA Details
| Gene Symbol: Tnfaip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182950.4 | NP_891995.1 | 5 | 786 | ||
| GenBank mRNA | AY336949.2 | - | 5 | 673 | ||
| BC128704.1 | - | 5 | 786 | 7459404 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnfaip1| Gene Symbol: Tnfaip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182950.4 | NP_891995.1 | 5 | 786 | ||
| GenBank mRNA | AY336949.2 | - | 5 | 673 | ||
| BC128704.1 | - | 5 | 786 | 7459404 | ||