siRNA Details
| Gene Symbol: Sarm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105817.1 | NP_001099287.1 | 2 | 626 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sarm1| Gene Symbol: Sarm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105817.1 | NP_001099287.1 | 2 | 626 | ||