siRNA Details
| Gene Symbol: Tp53i13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105818.1 | NP_001099288.1 | 6, 7 | 1122 | ||
| XM_006246902.2 | XP_006246964.1 | 5, 6 | 1149 | |||
| GenBank mRNA | BC158678.1 | - | 6, 7 | 1084 | 8372780 | |

Tp53i13
Genomic Map