siRNA Details
Gene Symbol: Spaca3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105820.1 | NP_001099290.1 | 3, 4 | 528 | ||
XM_006246983.2 | XP_006247045.1 | 2, 3 | 423 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Spaca3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105820.1 | NP_001099290.1 | 3, 4 | 528 | ||
XM_006246983.2 | XP_006247045.1 | 2, 3 | 423 |