siRNA Details
| Gene Symbol: Spaca3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105820.1 | NP_001099290.1 | 3 | 481 | ||
| XM_006246983.2 | XP_006247045.1 | 2 | 376 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spaca3| Gene Symbol: Spaca3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105820.1 | NP_001099290.1 | 3 | 481 | ||
| XM_006246983.2 | XP_006247045.1 | 2 | 376 | |||