siRNA Details
| Gene Symbol: Gas2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105823.2 | NP_001099293.2 | 4 | 796 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gas2l2| Gene Symbol: Gas2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105823.2 | NP_001099293.2 | 4 | 796 | ||