siRNA Details
Gene Symbol: Gas2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105823.2 | NP_001099293.2 | 6 | 2401 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gas2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105823.2 | NP_001099293.2 | 6 | 2401 |