siRNA Details
| Gene Symbol: Ppm1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105825.2 | NP_001099295.2 | 1 | 122 | ||
| GenBank mRNA | BC162058.1 | - | 1 | 122 | 7115026 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppm1d| Gene Symbol: Ppm1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105825.2 | NP_001099295.2 | 1 | 122 | ||
| GenBank mRNA | BC162058.1 | - | 1 | 122 | 7115026 | |