siRNA Details
| Gene Symbol: Sept4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011893.1 | NP_001011893.1 | 6 | 652 | ||
| XM_008767966.1 | XP_008766188.1 | 10 | 1832 | |||
| GenBank mRNA | AY208293.1 | - | 6 | 652 | ||
| AY208294.1 | - | 6 | 652 | |||
| BC088334.1 | - | 5 | 500 | |||

Sept4
Genomic Map