siRNA Details
| Gene Symbol: Mks1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034917.2 | NP_001030089.2 | 7 | 703 | ||
| GenBank mRNA | BC099806.1 | - | 7 | 701 | 7938236 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mks1| Gene Symbol: Mks1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034917.2 | NP_001030089.2 | 7 | 703 | ||
| GenBank mRNA | BC099806.1 | - | 7 | 701 | 7938236 | |