siRNA Details
| Gene Symbol: Wfikkn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247173.2 | XP_006247235.1 | 2 | 1716 | ||
| XM_220855.8 | XP_220855.1 | 2 | 2565 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wfikkn2| Gene Symbol: Wfikkn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247173.2 | XP_006247235.1 | 2 | 1716 | ||
| XM_220855.8 | XP_220855.1 | 2 | 2565 | |||