siRNA Details
| Gene Symbol: Mrpl27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105831.1 | NP_001099301.1 | 2 | 144 | ||
| GenBank mRNA | FQ212021.1 | - | 2 | 153 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrpl27| Gene Symbol: Mrpl27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105831.1 | NP_001099301.1 | 2 | 144 | ||
| GenBank mRNA | FQ212021.1 | - | 2 | 153 | ||