siRNA Details
| Gene Symbol: Eif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105837.1 | NP_001099307.1 | 4 | 981 | ||
| GenBank mRNA | BC159406.1 | - | 4 | 967 | 8361778 | |
| FQ212688.1 | - | 4 | 973 | |||
| FQ213689.1 | - | 4 | 830 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif1| Gene Symbol: Eif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105837.1 | NP_001099307.1 | 4 | 981 | ||
| GenBank mRNA | BC159406.1 | - | 4 | 967 | 8361778 | |
| FQ212688.1 | - | 4 | 973 | |||
| FQ213689.1 | - | 4 | 830 | |||