siRNA Details
Gene Symbol: Ptrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105841.1 | NP_001099311.1 | 2 | 1241 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ptrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105841.1 | NP_001099311.1 | 2 | 1241 |