siRNA Details
| Gene Symbol: Coasy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006954.1 | NP_001006955.1 | 3 | 956 | ||
| XM_006247276.2 | XP_006247338.1 | 3 | 1337 | |||
| XM_008767975.1 | XP_008766197.1 | 4 | 1158 | |||
| GenBank mRNA | BC083781.1 | - | 3 | 956 | 7096053 | |

Coasy
Genomic Map