siRNA Details
Gene Symbol: Coasy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006954.1 | NP_001006955.1 | 7 | 1467 | ||
XM_006247276.2 | XP_006247338.1 | 7 | 1848 | |||
XM_008767975.1 | XP_008766197.1 | 8 | 1669 | |||
GenBank mRNA | BC083781.1 | - | 7 | 1467 | 7096053 |
