siRNA Details
Gene Symbol: Psme3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011894.1 | NP_001011894.1 | 8 | 726 | ||
GenBank mRNA | BC089889.1 | - | 8 | 726 | 7374359 | |
FQ232375.1 | - | 6 | 411 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Psme3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011894.1 | NP_001011894.1 | 8 | 726 | ||
GenBank mRNA | BC089889.1 | - | 8 | 726 | 7374359 | |
FQ232375.1 | - | 6 | 411 |