siRNA Details
| Gene Symbol: Psme3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011894.1 | NP_001011894.1 | 6 | 557 | ||
| GenBank mRNA | BC089889.1 | - | 6 | 557 | 7374359 | |
| FQ232375.1 | - | 4 | 242 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psme3| Gene Symbol: Psme3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011894.1 | NP_001011894.1 | 6 | 557 | ||
| GenBank mRNA | BC089889.1 | - | 6 | 557 | 7374359 | |
| FQ232375.1 | - | 4 | 242 | |||