siRNA Details
| Gene Symbol: Tmem106a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024967.1 | NP_001020138.1 | 2 | 410 | ||
| XM_008767977.1 | XP_008766199.1 | 2 | 608 | |||
| XM_008767978.1 | XP_008766200.1 | 3 | 351 | |||
| XM_008767979.1 | XP_008766201.1 | 3 | 339 | |||
| GenBank mRNA | BC091170.1 | - | 2 | 410 | 7131003 | |

Tmem106a
Genomic Map