siRNA Details
| Gene Symbol: Rdm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105842.1 | NP_001099312.1 | 6 | 765 | ||
| GenBank mRNA | FQ215272.1 | - | 6 | 698 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rdm1| Gene Symbol: Rdm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105842.1 | NP_001099312.1 | 6 | 765 | ||
| GenBank mRNA | FQ215272.1 | - | 6 | 698 | ||