siRNA Details
Gene Symbol: Dhx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047844.1 | NP_001041309.1 | 22 | 3590 | ||
GenBank mRNA | AY383690.1 | - | 22 | 3590 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dhx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047844.1 | NP_001041309.1 | 22 | 3590 | ||
GenBank mRNA | AY383690.1 | - | 22 | 3590 |