siRNA Details
| Gene Symbol: Atxn7l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247470.2 | XP_006247532.1 | 1 | 447 | ||
| XM_006247472.2 | XP_006247534.1 | 1 | 446 | |||
| XM_008768223.1 | XP_008766445.1 | 1 | 449 | |||
| XM_008768224.1 | XP_008766446.1 | 1 | 449 | |||
| XM_008768225.1 | XP_008766447.1 | 1 | 448 | |||

Atxn7l3
Genomic Map