siRNA Details
| Gene Symbol: Fam171a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002727821.4 | XP_002727867.2 | 1, 2 | 268 | ||
| XM_006247473.2 | XP_006247535.1 | 1, 2 | 268 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam171a2| Gene Symbol: Fam171a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002727821.4 | XP_002727867.2 | 1, 2 | 268 | ||
| XM_006247473.2 | XP_006247535.1 | 1, 2 | 268 | |||