siRNA Details
| Gene Symbol: Higd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105844.1 | NP_001099314.1 | 2 | 487 | ||
| XM_008768263.1 | XP_008766485.1 | 3 | 325 | |||
| XM_008768264.1 | XP_008766486.1 | 3 | 210 | |||
| XM_008768265.1 | XP_008766487.1 | 3 | 353 | |||

Higd1b
Genomic Map