siRNA Details
| Gene Symbol: Eftud2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247544.2 | XP_006247606.1 | 10 | 898 | ||
| XM_006247545.2 | XP_006247607.1 | 10 | 892 | |||
| XM_213492.8 | XP_213492.3 | 11 | 988 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eftud2| Gene Symbol: Eftud2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247544.2 | XP_006247606.1 | 10 | 898 | ||
| XM_006247545.2 | XP_006247607.1 | 10 | 892 | |||
| XM_213492.8 | XP_213492.3 | 11 | 988 | |||