siRNA Details
| Gene Symbol: Eftud2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247544.2 | XP_006247606.1 | 22, 23 | 2354 | ||
| XM_006247545.2 | XP_006247607.1 | 22, 23 | 2348 | |||
| XM_213492.8 | XP_213492.3 | 23, 24 | 2444 | |||
| GenBank mRNA | BC089941.1 | - | 8, 9 | 797 | ||

Eftud2
Genomic Map