siRNA Details
| Gene Symbol: Eftud2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247544.2 | XP_006247606.1 | 11, 12 | 1059 | ||
| XM_006247545.2 | XP_006247607.1 | 11, 12 | 1053 | |||
| XM_213492.8 | XP_213492.3 | 12, 13 | 1149 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eftud2| Gene Symbol: Eftud2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247544.2 | XP_006247606.1 | 11, 12 | 1059 | ||
| XM_006247545.2 | XP_006247607.1 | 11, 12 | 1053 | |||
| XM_213492.8 | XP_213492.3 | 12, 13 | 1149 | |||