siRNA Details
| Gene Symbol: Fmnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247498.2 | XP_006247560.1 | 9 | 1158 | ||
| XM_006247499.2 | XP_006247561.1 | 9 | 1159 | |||
| XM_006247500.2 | XP_006247562.1 | 8 | 1140 | |||
| XM_006247501.2 | XP_006247563.1 | 8 | 1140 | |||
| XM_006247502.2 | XP_006247564.1 | 9 | 1128 | |||
| XM_008768266.1 | XP_008766488.1 | 7 | 1013 | |||
| XM_008768267.1 | XP_008766489.1 | 7 | 984 | |||

Fmnl1
Genomic Map