siRNA Details
| Gene Symbol: Lyzl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135833.1 | NP_001129305.2 | 5 | 470 | ||
| XM_006247503.2 | XP_006247565.1 | 5 | 1983 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lyzl6| Gene Symbol: Lyzl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135833.1 | NP_001129305.2 | 5 | 470 | ||
| XM_006247503.2 | XP_006247565.1 | 5 | 1983 | |||