siRNA Details
| Gene Symbol: Sppl2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105847.1 | NP_001099317.1 | 1 | 559 | ||
| XM_006247504.2 | XP_006247566.1 | 1 | 555 | |||
| GenBank mRNA | BC160911.1 | - | 1 | 528 | 7456885 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sppl2c| Gene Symbol: Sppl2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105847.1 | NP_001099317.1 | 1 | 559 | ||
| XM_006247504.2 | XP_006247566.1 | 1 | 555 | |||
| GenBank mRNA | BC160911.1 | - | 1 | 528 | 7456885 | |