siRNA Details
| Gene Symbol: Prr29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_213490.8 | XP_213490.2 | 5 | 2120 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr29| Gene Symbol: Prr29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_213490.8 | XP_213490.2 | 5 | 2120 | ||