siRNA Details
| Gene Symbol: Cep112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105849.1 | NP_001099319.1 | 14 | 1620 | ||
| XM_008768317.1 | XP_008766539.1 | 14 | 1654 | |||
| XM_008768318.1 | XP_008766540.1 | 14 | 1792 | |||
| XM_008768319.1 | XP_008766541.1 | 13 | 1510 | |||
| XM_008768320.1 | XP_008766542.1 | 14 | 1734 | |||
| XM_008768321.1 | XP_008766543.1 | 14 | 2228 | |||

Cep112
Genomic Map