siRNA Details
| Gene Symbol: Slc39a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013042.1 | NP_001013060.1 | 7 | 741 | ||
| XM_006247663.2 | XP_006247725.1 | 7 | 1060 | |||
| XM_006247664.2 | XP_006247726.1 | 7 | 1131 | |||
| XM_006247665.2 | XP_006247727.1 | 7 | 1039 | |||
| XM_006247668.2 | XP_006247730.1 | 7 | 827 | |||
| XM_006247670.2 | XP_006247732.1 | 7 | 1611 | |||
| XM_006247671.2 | XP_006247733.1 | 7 | 1210 | |||
| GenBank mRNA | BC062054.1 | - | 7 | 741 | 5598901 | |

Slc39a11
Genomic Map