siRNA Details
| Gene Symbol: TNP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005425.4 | NP_005416.1 | 1 | 416 | ||
| GenBank mRNA | BC096135.3 | - | 1 | 394 | 40002547 | |
| BC096136.3 | - | 1 | 394 | 40002550 | ||
| BC096137.1 | - | 1 | 394 | 40002553 | ||
| BX282422.1 | - | 1 | 416 | |||
| X63758.1 | - | 1 | 375 | |||
| X63759.1 | - | 1 | 643 | |||

TNP2
Genomic Map