siRNA Details
| Gene Symbol: Trim80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768412.1 | XP_008766634.1 | 8, 9 | 3327 | ||
| GenBank mRNA | BC091694.1 | - | 7, 8 | 1301 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim80| Gene Symbol: Trim80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768412.1 | XP_008766634.1 | 8, 9 | 3327 | ||
| GenBank mRNA | BC091694.1 | - | 7, 8 | 1301 | ||