siRNA Details
| Gene Symbol: Armc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127522.1 | NP_001120994.1 | 3 | 713 | ||
| GenBank mRNA | BC166454.1 | - | 3 | 687 | 9022599 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Armc7| Gene Symbol: Armc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127522.1 | NP_001120994.1 | 3 | 713 | ||
| GenBank mRNA | BC166454.1 | - | 3 | 687 | 9022599 | |