siRNA Details
| Gene Symbol: Galk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008282.1 | NP_001008283.1 | 2 | 265 | ||
| GenBank mRNA | BC085919.1 | - | 2 | 265 | 7108618 | |
| FQ219172.1 | - | 2 | 279 | |||
| FQ234739.1 | - | 2 | 238 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galk1| Gene Symbol: Galk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008282.1 | NP_001008283.1 | 2 | 265 | ||
| GenBank mRNA | BC085919.1 | - | 2 | 265 | 7108618 | |
| FQ219172.1 | - | 2 | 279 | |||
| FQ234739.1 | - | 2 | 238 | |||