siRNA Details
| Gene Symbol: Ubald2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_213532.7 | XP_213532.4 | 3 | 1192 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubald2| Gene Symbol: Ubald2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_213532.7 | XP_213532.4 | 3 | 1192 | ||