siRNA Details
| Gene Symbol: Rbfox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134498.2 | NP_001127970.1 | 9 | 949 | ||
| XM_008768343.1 | XP_008766565.1 | 9 | 2929 | |||
| XM_008768344.1 | XP_008766566.1 | 9 | 2928 | |||
| XM_008768345.1 | XP_008766567.1 | 9 | 2786 | |||
| XM_008768346.1 | XP_008766568.1 | 9 | 2785 | |||

Rbfox3
Genomic Map