siRNA Details
| Gene Symbol: Ccdc40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134688.1 | NP_001128160.1 | 4 | 1174 | ||
| XM_008768470.1 | XP_008766692.1 | 3 | 605 | |||
| XM_008768471.1 | XP_008766693.1 | 3 | 602 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc40| Gene Symbol: Ccdc40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134688.1 | NP_001128160.1 | 4 | 1174 | ||
| XM_008768470.1 | XP_008766692.1 | 3 | 605 | |||
| XM_008768471.1 | XP_008766693.1 | 3 | 602 | |||