siRNA Details
| Gene Symbol: Chmp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105856.1 | NP_001099326.1 | 3 | 266 | ||
| GenBank mRNA | BC070963.1 | - | 1 | 58 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chmp6| Gene Symbol: Chmp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105856.1 | NP_001099326.1 | 3 | 266 | ||
| GenBank mRNA | BC070963.1 | - | 1 | 58 | ||