siRNA Details
| Gene Symbol: Sectm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199082.2 | NP_954513.2 | 3 | 301 | ||
| XM_006247879.2 | XP_006247941.1 | 3 | 397 | |||
| XM_006247880.2 | XP_006247942.1 | 4 | 1221 | |||
| XM_006247881.2 | XP_006247943.1 | 3 | 938 | |||
| XM_006247882.2 | XP_006247944.1 | 3 | 394 | |||
| XM_006247884.2 | XP_006247946.1 | 4 | 1219 | |||
| GenBank mRNA | DY309177.1 | - | 2 | 389 | ||

Sectm1b
Genomic Map