siRNA Details
| Gene Symbol: Sectm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013043.1 | NP_001013061.1 | 5 | 679 | ||
| GenBank mRNA | BC078937.1 | - | 5 | 679 | 7108345 | |
| FQ225276.1 | - | 5 | 672 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sectm1a| Gene Symbol: Sectm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013043.1 | NP_001013061.1 | 5 | 679 | ||
| GenBank mRNA | BC078937.1 | - | 5 | 679 | 7108345 | |
| FQ225276.1 | - | 5 | 672 | |||