siRNA Details
| Gene Symbol: Mettl23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008283.1 | NP_001008284.1 | 4 | 541 | ||
| XM_006247798.1 | XP_006247860.1 | 4 | 527 | |||
| GenBank mRNA | BC086594.1 | - | 4 | 541 | 7310322 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mettl23| Gene Symbol: Mettl23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008283.1 | NP_001008284.1 | 4 | 541 | ||
| XM_006247798.1 | XP_006247860.1 | 4 | 527 | |||
| GenBank mRNA | BC086594.1 | - | 4 | 541 | 7310322 | |