siRNA Details
Gene Symbol: St6galnac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105859.1 | NP_001099329.1 | 7 | 1348 | ||
GenBank mRNA | AJ634458.1 | - | 7 | 1247 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: St6galnac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105859.1 | NP_001099329.1 | 7 | 1348 | ||
GenBank mRNA | AJ634458.1 | - | 7 | 1247 |