siRNA Details
| Gene Symbol: Yars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009627.1 | NP_001009627.1 | 1 | 757 | ||
| GenBank mRNA | BC088224.1 | - | 1 | 757 | 7322099 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Yars2| Gene Symbol: Yars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009627.1 | NP_001009627.1 | 1 | 757 | ||
| GenBank mRNA | BC088224.1 | - | 1 | 757 | 7322099 | |