siRNA Details
| Gene Symbol: Pkp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100499.1 | NP_001093969.1 | 1739 | |||
| GenBank mRNA | BC092602.1 | - | 3 | 346 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pkp2| Gene Symbol: Pkp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100499.1 | NP_001093969.1 | 1739 | |||
| GenBank mRNA | BC092602.1 | - | 3 | 346 | ||