siRNA Details
| Gene Symbol: Top3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105861.1 | NP_001099331.1 | 18 | 2245 | ||
| XM_006248671.2 | XP_006248733.1 | 17 | 2379 | |||
| XM_006248673.2 | XP_006248735.1 | 17 | 2191 | |||
| GenBank mRNA | FQ211004.1 | - | 17 | 2192 | ||
| FQ212059.1 | - | 16 | 2919 | |||

Top3b
Genomic Map